Aminoacetonitrile sulfate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Aminoacetonitrile hemisulfate, 2-Aminoacetonitrile hemisulphate, Glycinonitrile sulfate, Glycinonitrile sulphate, Cyanomethylamine sulphate, Cyanomethylamine sulfate

B22155-14 B22155-22 B22155-36
CAAAAB22155-14EA 76.31 CAD
CAAAAB22155-14 CAAAAB22155-22 CAAAAB22155-36
Aminoacetonitrile sulfate 97%
Aminoacetonitrile sulfate

Purity: 97% CAS No.: 5466-22-8 Molecular Formula: C4H8N8.H2SO4 Formula Weight: 210.21 Odor: Odorless Melting Point: ca 160° dec. Beilstein Reference: 3916094 UN No.: 3439 MDL No.: MFCD00012851

Formula: C₄H₁₀N₄O₄S
MW: 210.21 g/mol
Melting Pt: ca 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012851
CAS Number: 5466-22-8
EINECS: 226-772-0
UN: 3439
ADR: "6.1",III
Merck Index: 14,412

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR