Diethyl cyclobutane-1,1-dicarboxylate 95%
Supplier: Thermo Scientific Chemicals
Warning
|
B22140-09
B22140-18
CAAAB22140-09EA
51.67
CAD
CAAAB22140-09
CAAAB22140-18
Diethyl cyclobutane-1,1-dicarboxylate 95%
Diethyl cyclobutane-1,1-dicarboxylate
Purity: 95%
CAS No.: 3779-29-1
Molecular Formula: C10H16O4
Formula Weight: 200.24
Flash Point: 89°(192°F)
Refractive Index: 1.4360
Boiling Point: 96-98°/8mm
Beilstein Reference: 2050195
MDL No.: MFCD00019261
Formula:
C₁₀H₁₆O₄ Boiling Pt: 96…98 °C/8 mm Density: 1.050 Flash Pt: 89 °C(192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019261 CAS Number: 3779-29-1 EINECS: 223-239-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...