4-(Chloromethyl)benzonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22138-09
B22138-18
B22138-30
CAAAB22138-09EA
45.19
CAD
CAAAB22138-09
CAAAB22138-18
CAAAB22138-30
4-(Chloromethyl)benzonitrile 98+%
4-(Chloromethyl)benzonitrile
Purity: 98+%
CAS No.: 874-86-2
Molecular Formula: C8H6ClN
Formula Weight: 151.60
Melting Point: 77-80°
UN No.: 2923
MDL No.: MFCD00019761
Formula:
C₈H₆ClN Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019761 CAS Number: 874-86-2 EINECS: 212-869-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...