Penta-O-acetyl-ɑ-D-glucopyranose 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,2,3,4,6-Penta-O-acetyl-α-D-glycopyranose, ɑ-D-Glucose Pentaacetate, 1,2,3,4,6-Penta-O-Acetyl-A-D-Glycopyranose, α-D(+)-Glucose pentaacetate, (2R,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, Pentaacetyl-ɑ-D-glucose, α-D-Glucose pentaacetate

B22137-18 B22137-30
CAAAB22137-18EA 59.88 CAD
CAAAB22137-18 CAAAB22137-30
Penta-O-acetyl-ɑ-D-glucopyranose 99%
Penta-O-acetyl-alpha-D-glucopyranose

Purity: 99% CAS No.: 604-68-2 Molecular Formula: C16H22O11 Formula Weight: 390.34 Melting Point: 109-113° Beilstein Reference: 98852 Optical Rotation: +102° (c=5 in chloroform) MDL No.: MFCD00064071

Formula: C₁₆H₂₂O₁₁
MW: 390.34 g/mol
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00064071
CAS Number: 604-68-2
EINECS: 210-073-2

Order Now

Specification Test Results

Optical rotation +102° (c=5 in chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR