1-(Trifluoromethyl)vinyl acetate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22133-18
B22133-30
CAAAB22133-18EA
739.44
CAD
CAAAB22133-18
CAAAB22133-30
1-(Trifluoromethyl)vinyl acetate 97%
1-(Trifluoromethyl)vinyl acetate
Purity: 97%
CAS No.: 2247-91-8
Molecular Formula: C5H5F3O2
Formula Weight: 154.09
Storage Temperature: 2°C to 8°C
Flash Point: 2°(35°F)
Refractive Index: 1.3420
Odor: Pungent
Boiling Point: 86-88°
Beilstein Reference: 2078331
UN No.: 3272
MDL No.: MFCD00040844
Formula:
C₅H₅F₃O₂ Boiling Pt: 86…88 °C Density: 1.200 Flash Pt: 2 °C (35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040844 CAS Number: 2247-91-8 EINECS: 218-838-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...