Methyl-3-hydroxy-2-thiophenecarboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22129-03
B22129-06
B22129-14
CAAAB22129-03EA
84.69
CAD
CAAAB22129-03
CAAAB22129-06
CAAAB22129-14
Methyl-3-hydroxy-2-thiophenecarboxylate 97%
Methyl-3-hydroxy-2-thiophenecarboxylate
Purity: 97%
CAS No.: 5118-06-9
Molecular Formula: C6H6O3S
Formula Weight: 158.18
Melting Point: 44-48°
Boiling Point: 107-109°/13mm
MDL No.: MFCD00055642
Formula:
C₆H₆O₃S Boiling Pt: 107…109 °C/13 mm Melting Pt: 44…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055642 CAS Number: 5118-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...