Dowex® 50WX8 (H), Dowex® 50-100 mesh

Supplier: Thermo Scientific Chemicals
Danger

B22109-0E B22109-22 B22109-36
CAAAB22109-0EEA 2215.16 CAD
CAAAB22109-0E CAAAB22109-22 CAAAB22109-36
Dowex® 50WX8 (H), Dowex® 50-100 mesh
Dowex® 50WX8 (H)

Fieser: 1,511 4,266 5,355 6,302 9,256 18,194


CAS No.: 11119-67-8 MDL No.: MFCD00132726

Storage Temperature: Ambient
MDL Number: MFCD00132726
CAS Number: 11119-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR