4-(Benzyloxy)-3,5-dimethylbenzaldehyde 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Benzyloxy-3,5-dimethylbenzaldehyde

B22067-03 B22067-06
CAAAB22067-03EA 66.36 CAD
CAAAB22067-03 CAAAB22067-06
4-(Benzyloxy)-3,5-dimethylbenzaldehyde 95%
4-(Benzyloxy)-3,5-dimethylbenzaldehyd

Purity: 95% CAS No.: 144896-51-5 Molecular Formula: C16H16O2 Formula Weight: 240.30 Flash Point: 110°(230°F) Refractive Index: 1.5860 Boiling Point: 162-163°/1mm MDL No.: MFCD00800683

Formula: C₁₆H₁₆O₂
MW: 240.30 g/mol
Boiling Pt: 162…163 °C/1 mm
Density: 1.10
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00800683
CAS Number: 144896-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR