4-Bromo-3-chloro-3,4,4-trifluorobut-1-ene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22057-06
B22057-14
CAAAB22057-06EA
30.65
CAD
CAAAB22057-06
CAAAB22057-14
4-Bromo-3-chloro-3,4,4-trifluorobut-1-ene 97%
4-Bromo-3-chloro-3,4,4-trifluorobut-1-ene
Purity: 97%
CAS No.: 374-25-4
Molecular Formula: CH2=CHCFClCF2Br
Formula Weight: 223.43
Refractive Index: 1.4070
Boiling Point: 98-99°
Beilstein Reference: 1758700
MDL No.: MFCD00039225
Formula:
C₄H₃BrClF₃ Boiling Pt: 98…99 °C Density: 1.678 Storage Temperature: Ambient |
MDL Number:
MFCD00039225 CAS Number: 374-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...