Methyl-5-iodsalicylate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 2-hydroxy-5-iodobenzoate
B22048-09
B22048-18
CAAAB22048-09EA
116.45
CAD
CAAAB22048-09
CAAAB22048-18
CAAAB22048-30
Methyl-5-iodsalicylate 99%
Methyl 5-iodsalicylate
Purity: 99%
CAS No.: 4068-75-1
Molecular Formula: C8H7IO3
Formula Weight: 278.05
Melting Point: 75-78°
MDL No.: MFCD00016462
Formula:
C₈H₇IO₃ MW: 278.05 g/mol Melting Pt: 75…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016462 CAS Number: 4068-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...