tert-Butyl trichloroacetimidate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22039-06
B22039-14
CAAAAB22039-03EA
55.3
CAD
CAAAAB22039-03
CAAAB22039-06
CAAAB22039-14
tert-Butyl trichloroacetimidate 97%
tert-Butyl trichloroacetimidate
Fieser: 14,70
Purity: 97%
CAS No.: 98946-18-0
Molecular Formula: C6H10Cl3NO
Formula Weight: 218.51
Storage Temperature: 2°C to 8°C
Flash Point: 55°(131°F)
Physical Form: Liquid
Density: 1.222
Refractive Index: 1.4570
Sensitivity: Moisture Sensitive
Appearance: Colorless to pale yellow
Melting Point: 20-22°
Boiling Point: 64-66°/15mm
Beilstein Reference: 1770049
UN No.: 1993
MDL No.: MFCD00077410
Formula:
C₆H₁₀Cl₃NO Boiling Pt: 64…66 °C/15 mm Melting Pt: 20…22 °C Density: 1.222 Flash Pt: 55 °C(131 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00077410 CAS Number: 98946-18-0 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...