O-Methylisourea hemisulfate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22036-0B
B22036-18
B22036-30
CAAAB22036-0BEA
910.08
CAD
CAAAB22036-0B
CAAAB22036-18
CAAAB22036-30
O-Methylisourea hemisulfate 99%
O-Methylisourea hemisulfate
Purity: 99%
CAS No.: 52328-05-9
Molecular Formula: C2H6N2O.0.5H2SO4
Formula Weight: 123.12
Melting Point: 163-167°
MDL No.: MFCD00040594
Formula:
C₄H₁₄N₄O₆S Melting Pt: 163…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040594 CAS Number: 52328-05-9 EINECS: 257-851-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...