Mesitylenesulfonic acid sodium salt hemihydrate 98%

Supplier: Thermo Scientific Chemicals

B22029-14 B22029-22 B22029-36
CAAAB22029-14EA 94.17 CAD
CAAAB22029-14 CAAAB22029-22 CAAAB22029-36
Mesitylenesulfonic acid sodium salt hemihydrate 98%
Mesitylenesulfonic acid sodium salt hemihydrate

Purity: 98% CAS No.: 6148-75-0 Molecular Formula: C9H11NaO3S.0.5H2O Formula Weight: 231.26 (222.25anhy) Melting Point: 300° Beilstein Reference: 5784990 MDL No.: MFCD00067712

Formula: C₉H₁₁NaO₃S·¹/₂H₂O
MW: 231.26 g/mol (222.25 anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00067712
CAS Number: 6148-75-0
EINECS: 228-157-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR