1,4'-Bipiperidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Piperidinopiperidine

B22028-03 B22028-06 B22028-14
CAAAB22028-03EA 86.74 CAD
CAAAB22028-03 CAAAB22028-06 CAAAB22028-14
1,4'-Bipiperidine 99%
1,4'-Bipiperidine

Purity: 99% CAS No.: 4897-50-1 Molecular Formula: C10H20N2 Formula Weight: 168.28 Melting Point: 66-70° UN No.: 3259 MDL No.: MFCD00006475

Formula: C₁₀H₂₀N₂
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00006475
CAS Number: 4897-50-1
EINECS: 225-522-8
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR