2-Methyl-5-(methylthio)furan 99%

Supplier: Thermo Scientific Chemicals
Warning

B21998-04 B21998-09
CAAAB21998-04EA 79.95 CAD
CAAAB21998-04 CAAAB21998-09
2-Methyl-5-(methylthio)furan 99%
2-Methyl-5-(methylthio)furan

Purity: 99% CAS No.: 13678-59-6 Molecular Formula: C7H14OS Formula Weight: 128.19 Flash Point: 50°(122°F) Odor: Stench Boiling Point: 79-81°/50mm UN No.: 1993 MDL No.: MFCD01208018

Formula: C₆H₈OS
Boiling Pt: 79…81 °C/50 mm
Density: 1.063
Flash Pt: 50 °C(122 °F)
Storage Temperature: Ambient
MDL Number: MFCD01208018
CAS Number: 13678-59-6
EINECS: 237-170-2
UN: 1993
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR