4-Chloro-2,6-xylidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21996-06
B21996-14
CAAAB21996-06EA
98.28
CAD
CAAAB21996-06
CAAAB21996-14
4-Chloro-2,6-xylidine 98%
4-Chloro-2,6-xylidine
Purity: 98%
CAS No.: 24596-18-7
Molecular Formula: C8H10ClN
Formula Weight: 155.63
Melting Point: 45-50°
MDL No.: MFCD01076576
Formula:
C₈H₁₀ClN Melting Pt: 45…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076576 CAS Number: 24596-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...