BAPTA tetraethyl ester ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Tetraethyl 2,2',2'',2'''-(((ethane-1,2-diylbis(oxy))bis(2,1-phenylene))bis(azanetriyl))tetraacetate

B21971-03 B21971-06
CAAAB21971-03EA 181.7 CAD
CAAAB21971-03 CAAAB21971-06 CAAAB21971-14
BAPTA tetraethyl ester ≥98%
BAPTA tetraethyl ester

Purity: 98+% CAS No.: 73630-07-6 Molecular Formula: C30H40N2O10 Formula Weight: 588.66 Melting Point: 98-102° MDL No.: MFCD00532663

Formula: C₃₀H₄₀N₂O₁₀
MW: 588.66 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00532663
CAS Number: 73630-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR