2,5-Diaminohydroquinone dihydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21969-01
B21969-03
CAAAB21969-01EA
94.8
CAD
CAAAB21969-01
CAAAB21969-03
2,5-Diaminohydroquinone dihydrochloride 97%
2,5-Diaminohydroquinone dihydrochloride
Purity: 97%
CAS No.: 24171-03-7
Molecular Formula: C6H8N2O2.2HCl
Formula Weight: 213.06
Odor: Odorless
Melting Point:
Formula:
C₆H₁₀Cl₂N₂O₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239416 CAS Number: 24171-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...