3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenazopyridine hydrochloride, 3-Phenylazo-2,6-diaminopyridine hydrochloride

B21886-06 B21886-14
CAAAB21886-03EA 70.37 CAD
CAAAB21886-03 CAAAB21886-06 CAAAB21886-14
3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride ≥98%
3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride

Purity: 98+% CAS No.: 136-40-3 Molecular Formula: C11H11N5.HCl Formula Weight: 249.70 Melting Point: ca 240° dec. MDL No.: MFCD00035347

Formula: C11H12ClN5
MW: 249.70 g/mol
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00035347
CAS Number: 136-40-3
EINECS: 205-243-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR