2-(4-Methyl-5-thiazolyl)ethyl acetate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

B21867-06 B21867-14 B21867-22
CAAAB21867-06EA 40.45 CAD
CAAAB21867-06 CAAAB21867-14 CAAAB21867-22
2-(4-Methyl-5-thiazolyl)ethyl acetate ≥98%
2-(4-Methyl-5-thiazolyl)ethyl acetate

Purity: 98+% CAS No.: 656-53-1 Molecular Formula: C8H11NO2S Formula Weight: 185.24 Flash Point: 110°(230°F) Refractive Index: 1.5100 Boiling Point: 116-118°/6mm MDL No.: MFCD00005338

Formula: C₈H₁₁NO₂S
Boiling Pt: 116…118 °C/6 mm
Density: 1.147
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005338
CAS Number: 656-53-1
EINECS: 211-515-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR