N-Acetyl-DL-methionine 99%

Supplier: Thermo Scientific Chemicals

B21866-18 B21866-0B B21866-30
CAAAAB21866-18EA 33.02 CAD
CAAAAB21866-18 CAAAB21866-0B CAAAB21866-30
N-Acetyl-DL-methionine 99%
N-Acetyl-DL-methionine

Purity: 99% CAS No.: 1115-47-5 Molecular Formula: C7H13NO3S Formula Weight: 191.25 Storage Temperature: 2°C to 8°C Melting Point: 113-117° Beilstein Reference: 1725554 Merck Reference: 14,96 MDL No.: MFCD00008681

Formula: C₇H₁₃NO₃S
Melting Pt: 113…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008681
CAS Number: 1115-47-5
EINECS: 214-224-3
Merck Index: 14,96

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR