Methyl-2-methoxy-5-nitrobenzoate 98%
Supplier: Thermo Scientific Chemicals
B21863-06
B21863-14
CAAAB21863-06EA
150.1
CAD
CAAAB21863-06
CAAAB21863-14
Methyl-2-methoxy-5-nitrobenzoate 98%
Methyl 2-methoxy-5-nitrobenzoate
Purity: 98%
CAS No.: 34841-11-7
Molecular Formula: C9H9NO5
Formula Weight: 211.18
Melting Point: 99-102°
MDL No.: MFCD00673023
Formula:
C₉H₉NO₅ Melting Pt: 99…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673023 CAS Number: 34841-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...