2-Ethylacrolein 90% stabilized, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with hydroquinone (HQ) 50 ppm
B21839-06
B21839-14
B21839-22
CAAAB21839-06EA
122.13
CAD
CAAAB21839-06
CAAAB21839-14
CAAAB21839-22
2-Ethylacrolein 90% stabilized, tech.
2-Ethylacrolein
Stab. with 50ppm hydroquinone
Purity: 90%
CAS No.: 922-63-4
Molecular Formula: C5H8O
Formula Weight: 84.12
Storage Temperature: 2°C to 8°C
Flash Point: 1°(34°F)
Refractive Index: 1.4245
Odor: Pungent
Boiling Point: 92-93°
Beilstein Reference: 1098378
UN No.: 1992
MDL No.: MFCD00010129
Formula:
C₅H₈O Boiling Pt: 92…93 °C Density: 0.859 Flash Pt: 1 °C(34 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010129 CAS Number: 922-63-4 EINECS: 213-079-3 UN: 1992 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...