Sodium ethanesulfonate, (max. 2% H₂O) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21824-06
B21824-14
CAAAAB21824-03EA
43.12
CAD
CAAAAB21824-03
CAAAAB21824-06
CAAAAB21824-14
Sodium ethanesulfonate, (max. 2% H₂O) 98%
Sodium ethanesulfonate
Purity: 98%
CAS No.: 5324-47-0
Molecular Formula: CH3CH2SO3Na
Formula Weight: 132.11
Melting Point: ca 270°
MDL No.: MFCD00066498
Formula:
C₂H₆O₃S Melting Pt: ca 270 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066498 CAS Number: 5324-47-0 EINECS: 226-194-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...