9-(Trimethylsilyl)fluorene 99%

Supplier: Thermo Scientific Chemicals

B21816-03 B21816-06
CAAAB21816-03EA 189.6 CAD
CAAAB21816-03 CAAAB21816-06
9-(Trimethylsilyl)fluorene 99%
9-(Trimethylsilyl)fluorene

Purity: 99% CAS No.: 7385-10-6 Molecular Formula: C16H18Si Formula Weight: 238.40 Melting Point: 95-98° MDL No.: MFCD00092570

Formula: C₁₆H₁₈Si
MW: 238.40 g/mol
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00092570
CAS Number: 7385-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR