1,1,2-Trimethyl-1H-benz[e]indole 97%

Supplier: Thermo Scientific Chemicals
Warning

B21804-06 B21804-14
CAAAB21804-03EA 71.65 CAD
CAAAB21804-03 CAAAB21804-06 CAAAB21804-14
1,1,2-Trimethyl-1H-benz[e]indole 97%
1,1,2-Trimethyl-1H-benz[e]indole

Purity: 97% CAS No.: 41532-84-7 Molecular Formula: C15H15N Formula Weight: 209.29 Melting Point: 114-119° Beilstein Reference: 153709

Formula: C₁₅H₁₅N
Melting Pt: 114…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00082627
CAS Number: 41532-84-7
EINECS: 255-429-8
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR