Naphthalene-1-sulfonic acid hydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

B21782-09 B21782-18
CAAAB21782-09EA 108.55 CAD
CAAAB21782-09 CAAAB21782-18 CAAAB21782-30
Naphthalene-1-sulfonic acid hydrate 98%
Naphthalene-1-sulfonic acid hydrate

Purity: 98% CAS No.: 6036-48-2 Molecular Formula: C10H8O3S.xH2O Formula Weight: 208.23(anhy) Melting Point: 89-91°(dried) UN No.: 2585 MDL No.: MFCD00003987

Formula: C₁₀H₈O₃S·xH₂O
MW: 208.23 g/mol (anhy)
Melting Pt: 89…91 °C (dried)
Storage Temperature: Ambient
MDL Number: MFCD00003987
CAS Number: 6036-48-2
EINECS: 201-610-1
UN: 2585
ADR: 8 III
Merck Index: 14,6376

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR