Phenyl glycidyl ether 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Glycidyl phenyl ether
B21768-0B
B21768-18
B21768-30
CAAAB21768-0BEA
605.14
CAD
CAAAB21768-0B
CAAAB21768-18
CAAAB21768-30
Phenyl glycidyl ether 99%
Phenyl glycidyl ether
Purity: 99%
CAS No.: 122-60-1
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 114°(237°F)
Refractive Index: 1.5310
Odor: Sweet
Melting Point: 3.5°
Boiling Point: 244-246°
Beilstein Reference: 2744
MDL No.: MFCD00005133
Formula:
C₉H₁₀O₂ MW: 150.18 Boiling Pt: 244…246 °C Melting Pt: 3.5 °C Density: 1.110 Flash Pt: 114 °C(237 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005133 CAS Number: 122-60-1 EINECS: 204-557-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...