3,3'-Dimethyldiphenylamine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21765-03
B21765-06
CAAAB21765-03EA
111.07
CAD
CAAAB21765-03
CAAAB21765-06
3,3'-Dimethyldiphenylamine 98%
3,3'-Dimethyldiphenylamine
Purity: 98%
CAS No.: 626-13-1
Molecular Formula: C14H15N
Formula Weight: 197.28
Flash Point:
Formula:
C₁₄H₁₅N Boiling Pt: 200 °C/24 mm Density: 1.04 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00059315 CAS Number: 626-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...