1,4-Oxazepane hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21749-03
B21749-06
CAAAB21749-03EA
492.96
CAD
CAAAB21749-03
CAAAB21749-06
1,4-Oxazepane hydrochloride 98%
1,4-Oxazepane hydrochloride
Purity: 98%
CAS No.: 178312-62-4
Molecular Formula: C5H11NO.HCl
Formula Weight: 137.61
Melting Point: 158-164°
MDL No.: MFCD02683070
Formula:
C₅H₁₂ClNO Melting Pt: 158…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683070 CAS Number: 178312-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...