4-Nitrobenzylamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Nitrophenyl)methanamine hydrochloride

B21748-03 B21748-06 B21748-14
CAAAB21748-03EA 69.52 CAD
CAAAB21748-03 CAAAB21748-06 CAAAB21748-14
4-Nitrobenzylamine hydrochloride 97%
4-Nitrobenzylamine hydrochloride

Purity: 97% CAS No.: 18600-42-5 Molecular Formula: C7H8N2O2.HCl Formula Weight: 188.62 Melting Point: 255-257° dec. Beilstein Reference: 3629994

Formula: C₇H₉ClN₂O₂
MW: 188.62 g/mol
Melting Pt: 255…257 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012863
CAS Number: 18600-42-5
EINECS: 242-441-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR