trans-4-(Aminomethyl)cyclohexanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: HAKU, AMCA, TAMCHA, trans-4-(aminomethyl)cyclohexane-1-carboxylic acid, Tranexamic acid, AMCHA

B21742-09 B21742-18 B21742-30
CAAAB21742-09EA 77.26 CAD
CAAAB21742-09 CAAAB21742-18 CAAAB21742-30
trans-4-(Aminomethyl)cyclohexanecarboxylic acid 97%
trans-4-(Aminomethyl)cyclohexanecarboxylic acid

Inhibits plasmin-induced fibrinolysis


Purity: 97% CAS No.: 1197-18-8 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Melting Point: 300° Beilstein Reference: 2207452 Merck Reference: 14,9569 MDL No.: MFCD00001466

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00001466
CAS Number: 1197-18-8
EINECS: 214-818-2
Merck Index: 14,9569

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR