Malonaldehydebis(phenylimine) monohydrochloride ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: N1,N3-Diphenylpropane-1,3-diimine hydrochloride

B21721-14 B21721-22
CAAAB21721-14EA 103.65 CAD
CAAAB21721-14 CAAAB21721-22
Malonaldehydebis(phenylimine) monohydrochloride ≥98%
Malonaldehydebis(phenylimine) monohydrochloride

Purity: 97+% CAS No.: 123071-42-1 Molecular Formula: C15H14N2.HCl Formula Weight: 258.75 Melting Point: ca 225° dec. MDL No.: MFCD00054634

Formula: C₁₅H₁₄N₂.HCl
MW: 258.75 g/mol
Melting Pt: ca 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00054634
CAS Number: 123071-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR