4-Methyl-5-vinylthiazole ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21719-06
B21719-14
B21719-22
CAAAB21719-06EA
55.62
CAD
CAAAB21719-06
CAAAB21719-14
CAAAB21719-22
4-Methyl-5-vinylthiazole ≥98%
4-Methyl-5-vinylthiazole
Purity: 98+%
CAS No.: 1759-28-0
Molecular Formula: C6H7NS
Formula Weight: 125.19
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Refractive Index: 1.5680
Odor: Pleasant
Melting Point: -15°
Boiling Point: 67-68°/11mm
Beilstein Reference: 107867
MDL No.: MFCD00005337
Formula:
C₆H₇NS Boiling Pt: 67…68 °C/11 mm Melting Pt: –15 °C Density: 1.095 Flash Pt: 70 °C(158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005337 CAS Number: 1759-28-0 EINECS: 217-160-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...