Direct red 80

Supplier: Thermo Scientific Chemicals

B21693-06 B21693-14 B21693-22
CAAAB21693-06EA 64.46 CAD
CAAAB21693-06 CAAAB21693-14 CAAAB21693-22
Direct red 80
Direct Red 80

CAS No.: 2610-10-8 Molecular Formula: C45H26N10Na6O21S6 Formula Weight: 1373.09 MDL No.: MFCD00054389

Formula: C₄₅H₂₆N₁₀Na₆O₂₁S₆
Storage Temperature: Ambient
MDL Number: MFCD00054389
CAS Number: 2610-10-8
EINECS: 220-027-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR