N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Thr(Bzl)-OH, (2S,3R)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)butanoic acid, N-Boc-O-benzyl-L-threonine, BOC-O-Benzyl-L-threonine

B21677-03 B21677-06
CAAAB21677-03EA 54.35 CAD
CAAAB21677-03 CAAAB21677-06
N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine 99%
N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine

Purity: 99% CAS No.: 15260-10-3 Molecular Formula: C16H23NO5 Formula Weight: 309.36 Melting Point: 115-116° MDL No.: MFCD00066062

Formula: C₁₆H₂₃NO₅
MW: 309.36 g/mol
Melting Pt: 115…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00066062
CAS Number: 15260-10-3
EINECS: 239-304-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR