N-Carbobenzoxy-L-proline ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Z-Pro-OH, Z-L-Prolin, N-Cbz-L-proline, N-Carbobenzyloxy-L-proline, N-Benzyloxycarbonyl-L-proline
B21672-06
B21672-18
B21672-30
CAAAAB21672-06EA
53.25
CAD
CAAAAB21672-06
CAAAB21672-18
CAAAB21672-30
N-Carbobenzoxy-L-proline ≥98%
N-Carbobenzoxy-L-proline
Purity: 98+%
CAS No.: 1148-11-4
Molecular Formula: C13H15NO4
Formula Weight: 249.27
Melting Point: 75-77°
Beilstein Reference: 88579
Optical Rotation: -60° (c=1 in acetic acid)
MDL No.: MFCD00003170
Formula:
C₁₃H₁₅NO₄ MW: 249.27 g/mol Melting Pt: 75…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003170 CAS Number: 1148-11-4 EINECS: 214-557-4 |
Specification Test Results
Optical rotation | -60° (c=1 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...