15-Hydroxypentadecanoic acid 99+%

Supplier: Thermo Scientific Chemicals

B21670-03 B21670-06
CAAAB21670-03EA 101.59 CAD
CAAAB21670-03 CAAAB21670-06
15-Hydroxypentadecanoic acid 99+%
15-Hydroxypentadecanoic acid

Purity: 99+% CAS No.: 4617-33-8 Molecular Formula: C15H30O3 Formula Weight: 258.40 Melting Point: 84-88° Beilstein Reference: 1240100

Formula: C₁₅H₃₀O₃
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00039535
CAS Number: 4617-33-8
EINECS: 225-026-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR