Tartronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21640-02
B21640-77
CAAAB21640-02EA
341.28
CAD
CAAAB21640-02
CAAAB21640-77
Tartronic acid 98%
Tartronic acid
Purity: 98%
CAS No.: 80-69-3
Molecular Formula: C3H4O5
Formula Weight: 120.06
Physical Form: Powder
Appearance: White
Melting Point: ca 151° dec.
Beilstein Reference: 1209791
Merck Reference: 14,9073
MDL No.: MFCD00004237
Formula:
C₃H₄O₅ Melting Pt: ca 151 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004237 CAS Number: 80-69-3 EINECS: 201-301-1 Merck Index: 14,9073 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...