Methyl-4-acetylbenzoate 99%
Supplier: Thermo Scientific Chemicals
B21626-04
B21626-09
CAAAB21626-04EA
108.23
CAD
CAAAB21626-04
CAAAB21626-09
Methyl-4-acetylbenzoate 99%
Methyl 4-acetylbenzoate
Purity: 99%
CAS No.: 3609-53-8
Molecular Formula: C10H10O3
Formula Weight: 178.19
Melting Point: 92-95°
MDL No.: MFCD00216474
Formula:
C₁₀H₁₀O₃ Melting Pt: 92…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216474 CAS Number: 3609-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...