Methyl trifluoropyruvate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21589-06
B21589-14
CAAAB21589-03EA
51.27
CAD
CAAAB21589-03
CAAAB21589-06
CAAAB21589-14
Methyl trifluoropyruvate 97%
Methyl trifluoropyruvate
Purity: 97%
CAS No.: 13089-11-7
Molecular Formula: C4H3F3O3
Formula Weight: 156.06
Flash Point: 26°(79°F)
Refractive Index: 1.3320
Boiling Point: 84-86°
Beilstein Reference: 1863490
UN No.: 3272
MDL No.: MFCD00114936
Formula:
C₄H₃F₃O₃ Boiling Pt: 84…86 °C Density: 1.52 Flash Pt: 26 °C (79 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00114936 CAS Number: 13089-11-7 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...