L(+)-Lyxose 99%
Supplier: Thermo Scientific Chemicals
B21583-03
B21583-06
CAAAB21583-03EA
125.61
CAD
CAAAB21583-03
CAAAB21583-06
L(+)-Lyxose 99%
L(+)-Lyxose
Purity: 99%
CAS No.: 1949-78-6
Molecular Formula: C5H10O5
Formula Weight: 150.13
Melting Point: 108-112°
MDL No.: MFCD00064111
Formula:
C₅H₁₀O₅ Melting Pt: 108…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064111 CAS Number: 1949-78-6 EINECS: 217-763-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...