Nonafluorovaleric acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21567-06
B21567-14
CAAAB21567-06EA
111.39
CAD
CAAAB21567-06
CAAAB21567-14
Nonafluorovaleric acid 97%
Nonafluorovaleric acid
Purity: 97%
CAS No.: 2706-90-3
Molecular Formula: CF3(CF2)3CO2H
Formula Weight: 264.06
Refractive Index: 1.2940
Boiling Point: 140°
Beilstein Reference: 1800087
UN No.: 3265
MDL No.: MFCD00040211
Formula:
C₅HF₉O₂ Boiling Pt: 140 °C Density: 1.713 Storage Temperature: Ambient |
MDL Number:
MFCD00040211 CAS Number: 2706-90-3 EINECS: 220-300-7 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...