4,4'-Anisoin 97%
Supplier: Thermo Scientific Chemicals
B21559-14
B21559-36
CAAAB21559-14EA
82.48
CAD
CAAAB21559-14
CAAAB21559-22
CAAAB21559-36
4,4'-Anisoin 97%
4,4'-Anisoin
Purity: 97%
CAS No.: 119-52-8
Molecular Formula: C16H16O4
Formula Weight: 272.30
Melting Point: 110-114°
Beilstein Reference: 3211892
Formula:
C₁₆H₁₆O₄ Melting Pt: 110…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008411 CAS Number: 119-52-8 EINECS: 204-330-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...