4-(Dimethylamino)cinnamic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-(Dimethylamino)cinnamic Acid

B21549-03 B21549-06
CAAAB21549-03EA 91.96 CAD
CAAAB21549-03 CAAAB21549-06
4-(Dimethylamino)cinnamic acid 99%
4-(Dimethylamino)cinnamic acid

Purity: 99% CAS No.: 1552-96-1 Molecular Formula: C11H13NO2 Formula Weight: 191.23 Melting Point: ca 222° dec. Beilstein Reference: 1368533

Formula: C₁₁H₁₃NO₂
MW: 191.23 g/mol
Melting Pt: ∼ 222 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004397
CAS Number: 1552-96-1
EINECS: 216-299-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR