N-(Chloroacetyl)morpholine 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21548-03
B21548-06
CAAAB21548-03EA
51.82
CAD
CAAAB21548-03
CAAAB21548-06
N-(Chloroacetyl)morpholine 97+%
N-(Chloroacetyl)morpholine
Purity: 97+%
CAS No.: 1440-61-5
Molecular Formula: C6H10ClNO2
Formula Weight: 163.61
Melting Point: 27-30°
UN No.: 3261
MDL No.: MFCD00721939
Formula:
C₆H₁₀ClNO₂ Melting Pt: 27…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00721939 CAS Number: 1440-61-5 EINECS: 215-880-3 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...