Diethyl oxalacetate sodium salt 95%

Supplier: Thermo Scientific Chemicals
Warning

B21486-22 B21486-36
CAAAB21486-22EA 82.32 CAD
CAAAB21486-22 CAAAB21486-36
Diethyl oxalacetate sodium salt 95%
Diethyl oxalacetate sodium salt

Purity: 95% CAS No.: 40876-98-0 Molecular Formula: C8H11NaO5 Formula Weight: 210.16 Melting Point: ca 185° Beilstein Reference: 4279609 MDL No.: MFCD00035571

Formula: C₈H₁₂O₅
Melting Pt: ca 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00035571
CAS Number: 40876-98-0
EINECS: 255-122-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR