Calmagite indicator

Supplier: Thermo Scientific Chemicals
Warning

B21482-06 B21482-14 B21482-22
CAAAB21482-06EA 55.14 CAD
CAAAB21482-06 CAAAB21482-14 CAAAB21482-22
Calmagite indicator
Calmagite

CAS No.: 3147-14-6 Molecular Formula: C17H14N2O5S Formula Weight: 358.37 Melting Point: 330° Beilstein Reference: 708596 Merck Reference: 14,1718 MDL No.: MFCD00011656

Formula: C₁₇H₁₄N₂O₅S
MW: 358.37
Melting Pt: >330 °C
Storage Temperature: Ambient
MDL Number: MFCD00011656
CAS Number: 3147-14-6
EINECS: 221-563-0
Merck Index: 14,1718

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR