1,4-Bis(2-hydroxyisopropyl)benzene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Bis(α-hydroxyisopropyl)benzene, 1,4-Bis(1-methyl-1-hydroxyethyl)benzene, 1,4-Bis(alpha-hydroxyisopropyl)benzene, alpha,alpha'-Dihydroxy-1,4-diisopropylbenzene, 2,2'-(p-Phenylene)di-2-propanol, 2,2'-(1,4-Phenylene)bis(propan-2-ol), ^a,^a,^a',^a'-Tetramethyl-1,4-benzenedimethanol, alpha,alpha,alpha',alpha'-Tetramethyl-1,4-benzenedimethanol, α,α,α',α'-Tetramethyl-1,4-benzenedimethanol, α,α'-Dihydroxy-1,4-diisopropylbenzene, 1,4-Bis(2-hydroxyisopropyl)benzene

B21472-03 B21472-06
CAAAB21472-03EA 48.51 CAD
CAAAB21472-03 CAAAB21472-06
1,4-Bis(2-hydroxyisopropyl)benzene 97%
1,4-Bis(2-hydroxyisopropyl)benzene

Purity: 97% CAS No.: 2948-46-1 Molecular Formula: C12H18O2 Formula Weight: 194.28 Melting Point: 142-146° Boiling Point: 167-168°/12mm MDL No.: MFCD00009827

Formula: C₁₂H₁₈O₂
MW: 194.28 g/mol
Boiling Pt: 167…168 °C/12 mm
Melting Pt: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00009827
CAS Number: 2948-46-1
EINECS: 220-964-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR