Pseudothiohydantoin 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-aminothiazol-4(5H)-one, 2-Amino-4,5-dihydro-1,3-thiazol-4-one

B21461-06 B21461-14 B21461-22
CAAAB21461-06EA 65.25 CAD
CAAAB21461-06 CAAAB21461-14 CAAAB21461-22
Pseudothiohydantoin 97%
Pseudothiohydantoin

Purity: 97% CAS No.: 556-90-1 Molecular Formula: C3H4N2OS Formula Weight: 116.14 Melting Point: ca 240° dec. MDL No.: MFCD00003186

Formula: C₃H₄N₂OS
MW: 116.14 g/mol
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003186
CAS Number: 556-90-1
EINECS: 209-145-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR